Learn More
Sino Biological Recombinant Mouse ANGPTL3/Angiopoietin-like 3 Protein (His Tag)
Supplier:  Sino Biological 50003M08B200
Encompass_Preferred
A DNA sequence encoding the mouse ANGPTL3 (NP_038941.1) (Met1-Thr455) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3562