Learn More
Sino Biological Mouse ANGPTL4 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50356RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse ANGPTL4 / angiopoietin-like protein 4 (rM ANGPTL4 / angiopoietin-like protein 4; Catalog#50356-M07H; Q9Z1P8; Lys167-Ser410). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-1274