Learn More
Biomatik Corporation Mouse Ankyrin Repeat Domain Protein 1 (ANKRD1) ELISA Kit
Supplier:  Biomatik Corporation EKU02420
Encompass_Preferred
Mouse Ankyrin Repeat Domain Protein 1 (ANKRD1) ELISA Kit for detecting ANKRD1 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.052ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9CR42. Gene ID: 107765. Target Synonyms: CARP; ALRP; C-193; CVARP; MCARP; Ankyrin Repeat Domain 1 (Cardiac Muscle) ; Cardiac ankyrin repeat protein; Cytokine-inducible gene C-193; Cytokine-inducible nuclear protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4867