Learn More
Sino Biological Anti-CD13 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50001RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse ANPEP metalloprotease domain (rM ANPEP; Catalog#50001-M08H; Lys 66-Ser 966; NP_032512.2). ANPEP specific IgG was purified by mouse ANPEP affinity chromatography.
Catalog No. 50-255-3565