Learn More
Immunology Consultants Monoclonal Mouse anti-6xHIS Tag Antibody - Clone 7B8, 0.1 mg
SDP
Supplier:  Immunology Consultants MHIS45AZ
Encompass_Preferred
This mouse IgG2B is purified using salt fractionation procedures and Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with 6xHIS as determined by ELISA and western blot. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-196-2025