Learn More
Immunology Consultants Monoclonal Mouse anti-Alexa Fluor 647 Antibody, 1.0 mg
SDP
Supplier:  Immunology Consultants M64765A400
Encompass_Preferred
This mouse IgG is purified using salt fractionation procedures and Protein A chromatography. This antibody reacts with Alexa Fluor 647 as determined by ELISA, western blot, and lateral flow. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-196-2146