Learn More
Mouse Anti-Armenian and Syrian Hamster IgG1, Clone: HIG-632, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
Based on ELISA, the HIG-632 antibody reacts specifically with Armenian and Syrian hamster IgG1 monoclonal antibodies. The HIG-632 mAb does not react with other hamster IgG groups or hamster IgM.
Host Species: Mouse
Clone: HIG-632
Isotype: IgG2b κ
Species Reactivity: Hamster
Immunogen: Pooled Armenian Hamster IgG mAb
ELISA
Specifications
Specifications
| Antigen | Mouse Anti-Armenian and Syrian Hamster IgG1 |
| Classification | Monoclonal |
| Clone | HIG-632 |
| Concentration | 0.5mg/mL |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.