Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Bovine IgG2 Heavy Chain Secondary Antibody (IL-A73), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB555866
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB555866 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB555866 Supplier R&D Systems Supplier No. NBP268417C

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

IgG2 Heavy Chain Monoclonal specifically detects IgG2 Heavy Chain in Bovine samples. It is validated for ELISA, Flow Cytometry, Immunoprecipitation.

Specifications

Antigen IgG2 Heavy Chain
Applications ELISA, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone IL-A73
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Purified bovine IgG2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.