Learn More
Mouse anti-Bovine IgM (H+L) Secondary Antibody (IL-A30), DyLight 405, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgM (H+L) |
| Applications | ELISA, Flow Cytometry, Immunoprecipitation, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Clone | IL-A30 |
| Conjugate | DyLight 405 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Bovine peripheral blood mononuclear cells depleted of T lymphocytes |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.