Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Bovine IgM (H+L) Secondary Antibody (IL-A30), Janelia Fluor 549, Novus Biologicals™
SDP

Catalog No. NB554812
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB554812 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB554812 Supplier R&D Systems Supplier No. NBP268419JF549

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

IgM (H+L) Monoclonal specifically detects IgM (H+L) in Bovine samples. It is validated for ELISA, Flow Cytometry, Immunoprecipitation, Radioimmune Assays (RIA).

Specifications

Antigen IgM (H+L)
Applications ELISA, Flow Cytometry, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone IL-A30
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Bovine peripheral blood mononuclear cells depleted of T lymphocytes
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.