Learn More
Mouse anti-Canine IgA/IgG/IgM (H+L) Secondary Antibody (K99 2E3), PE, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgA/IgG/IgM (H+L) |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | K99 2E3 |
| Conjugate | PE |
| Formulation | PBS |
| Host Species | Mouse |
| Immunogen | Canine immunoglobulin |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.