Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Canine IgG2 Heavy Chain Secondary Antibody (CA4F1), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB554687
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB554687 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB554687 Supplier R&D Systems Supplier No. NBP268409UV

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

IgG2 Heavy Chain Monoclonal specifically detects IgG2 Heavy Chain in Canine samples. It is validated for ELISA, Flow Cytometry, Western Blot.

Specifications

Antigen IgG2 Heavy Chain
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone CA4F1
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Canine IgG
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.