Learn More
Immunology Consultants anti-Cat IgE Antibody (Mouse - Clone #2C1) - Monoclonal
SDP
Supplier:  Immunology Consultants ME20A2C1
Encompass_Preferred
This monoclonal antibody provides sensitive, specific detection of feline IgE, supporting ELISA, Western blotting, and allergy‑related assay development. Clone 2C1 delivers strong affinity and low background, enabling accurate measurement of IgE‑mediated immune responses in cats. It is well suited for veterinary diagnostics, hypersensitivity research, and studies examining allergic disease mechanisms. ME‑20A‑2C1 offers consistent lot‑to‑lot performance, ensuring reliable IgE quantification across serum, plasma, and purified immunoglobulin samples
Catalog No. 50-224-8857