Learn More
Immunology Consultants anti-Cat IgE Antibody (Mouse - Clone #ME1) - Monoclonal
SDP
Catalog No. 502248856
Encompass_Preferred
Clone ME1 targets a distinct epitope on feline IgE, providing complementary detection capabilities to Clone 2C1. It supports sensitive measurement of IgE in ELISA, Western blotting, and antigen‑capture assays, offering strong affinity and minimal nonspecific binding. ME‑20A‑ME1 is ideal for veterinary allergy research, diagnostic assay development, and monitoring IgE‑associated immune responses. Its reproducible performance ensures dependable results across diverse sample types.
This item is not returnable. View return policy
Catalog No. 50-224-8856 Supplier Immunology Consultants Supplier No. ME20AME1
Clone ME1 targets a distinct epitope on feline IgE, providing complementary detection capabilities to Clone 2C1. It supports sensitive measurement of IgE in ELISA, Western blotting, and antigen‑capture assays, offering strong affinity and minimal nonspecific binding. ME‑20A‑ME1 is ideal for veterinary allergy research, diagnostic assay development, and monitoring IgE‑associated immune responses. Its reproducible performance ensures dependable results across diverse sample types.