Learn More
Biolegend Purified anti-mouse CD28 (Maxpar Ready)
Supplier:  Biolegend 102119
Encompass_Preferred
Purified anti-mouse CD28 (Maxpar Ready) [37.51]; Isotype: Syrian Hamster IgG; Reactivity: Mouse; Apps: FC, CyTOF; Size: 100 ug
Catalog No. NC1365727