Learn More
Biolegend Purified anti-mouse CD80 (Maxpar Ready)
Supplier:  Biolegend 104735
Encompass_Preferred
Purified anti-mouse CD80 (Maxpar Ready) [16-10A1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Cross-Reactivity: Dog (Canine); Apps: FC, CyTOF; Size: 100 ug
Catalog No. NC1365726