Learn More
Chondrex Inc MOUSE ANTI-DNA IGG ASSAY, 1 KT
Catalog No. 501526760
Encompass_Preferred
ELISA kit for assaying mouse anti-double stranded DNA (dsDNA) IgG antibodies in mouse serum and plasma samples. Kit contains a 96-well ELISA plate, as well as all reagents needed to complete the assay. Standard Range: 50 ng/mL to 0.78 ng/mL. Working Time: 4.5 hours.
Catalog No. 50-152-6760 Supplier Chondrex Inc Supplier No. 3031
ELISA kit for assaying mouse anti-double stranded DNA (dsDNA) IgG antibodies in mouse serum and plasma samples. Kit contains a 96-well ELISA plate, as well as all reagents needed to complete the assay. Standard Range: 50 ng/mL to 0.78 ng/mL. Working Time: 4.5 hours.