Learn More
Immunology Consultants anti-Dog CRP Antibody (Mouse - Clone #14A2) - Monoclonal
SDP
Catalog No. 502248806
Encompass_Preferred
This monoclonal antibody provides sensitive, specific detection of canine C‑reactive protein (CRP), a major acute‑phase biomarker. Clone 14A2 performs reliably in ELISA, Western blotting, and antigen‑capture assays, offering strong affinity and low background. It is ideal for veterinary diagnostics, inflammation research, and monitoring CRP levels in canine serum or plasma. MCRP‑40A‑14A2 delivers consistent, high‑clarity performance across repeated assays.
This item is not returnable. View return policy
Catalog No. 50-224-8806 Supplier Immunology Consultants Supplier No. MCRP40A14A2
This monoclonal antibody provides sensitive, specific detection of canine C‑reactive protein (CRP), a major acute‑phase biomarker. Clone 14A2 performs reliably in ELISA, Western blotting, and antigen‑capture assays, offering strong affinity and low background. It is ideal for veterinary diagnostics, inflammation research, and monitoring CRP levels in canine serum or plasma. MCRP‑40A‑14A2 delivers consistent, high‑clarity performance across repeated assays.