Learn More
Immunology Consultants anti-Dog CRP Antibody (Mouse - Clone #14H5) - Monoclonal
SDP
Catalog No. 502248807
Encompass_Preferred
This monoclonal antibody recognizes a distinct canine CRP epitope, enabling sensitive detection in ELISA, Western blotting, and antigen‑capture assays. Clone 14H5 offers strong affinity, low nonspecific binding, and consistent lot‑to‑lot performance. It is well suited for veterinary diagnostics, inflammation research, and studies evaluating CRP as a biomarker of disease progression or therapeutic response.
This item is not returnable. View return policy
Catalog No. 50-224-8807 Supplier Immunology Consultants Supplier No. MCRP40A14H5
This monoclonal antibody recognizes a distinct canine CRP epitope, enabling sensitive detection in ELISA, Western blotting, and antigen‑capture assays. Clone 14H5 offers strong affinity, low nonspecific binding, and consistent lot‑to‑lot performance. It is well suited for veterinary diagnostics, inflammation research, and studies evaluating CRP as a biomarker of disease progression or therapeutic response.