Learn More
Immunology Consultants anti-Dog IgE Antibody (Mouse - Clone #E671) - Monoclonal
SDP
Catalog No. 502248860
Encompass_Preferred
This monoclonal antibody targets a distinct canine IgE epitope, providing sensitive, specific detection in ELISA and Western blotting. Clone E671 offers strong affinity, low background, and consistent lot‑to‑lot performance. It is well suited for veterinary immunology research, allergy diagnostics, and studies examining IgE‑mediated immune responses in dogs. ME‑40A‑E671 delivers reliable, high‑clarity detection across varied sample types.
This item is not returnable. View return policy
Catalog No. 50-224-8860 Supplier Immunology Consultants Supplier No. ME40AE671
This monoclonal antibody targets a distinct canine IgE epitope, providing sensitive, specific detection in ELISA and Western blotting. Clone E671 offers strong affinity, low background, and consistent lot‑to‑lot performance. It is well suited for veterinary immunology research, allergy diagnostics, and studies examining IgE‑mediated immune responses in dogs. ME‑40A‑E671 delivers reliable, high‑clarity detection across varied sample types.