Learn More
Columbia Biosciences Corp Mouse anti-DYKDDDDK IgG: SureLight™ APC;100ug
Supplier:  Columbia Biosciences Corp D31718100
Encompass_Preferred
Activity of Mouse Anti-DYKDDDDK MAbFlow Cytometry analysis of transfected HEK293 cells. Fixed and permeabilized cells stained with 1 µg/ml of Mouse anti-DYKDDDDK: SureLight APC. • Shaded histogram – HEK293 cells, expressing DYKDDDDK-tagged protein• Open histogram – mock transfected control HEK293 cells
Catalog No. 50-921-462