Learn More
Creative Biolabs MOUSE ANTI-EHV4 ORF16
SDP
Supplier:  Creative Biolabs VRH03088F
Encompass
Mouse Anti-EHV4 ORF16 (FSTAHSSAKSRRVGQLGVIP) Monoclonal Antibody (03088YF)This mouse monoclonal antibody 03088YF is generated from premade hybridoma library, and specific for Equid alphaherpesvirus 4 ORF16 (AA 157-176): FSTAHSSAKSRRVGQLGVIP. For any additional requirments, please specify and submit an inquiry for assistance.
Catalog No. NC2235663
Add to Cart
Request a Quote