Learn More
Creative Biolabs MOUSE ANTI-EHV4 ORF74 1MG
SDP
Supplier:  Creative Biolabs VRH03091F
Encompass
Mouse Anti-EHV4 ORF74 (KKPPTLPRVHVKTPPPILVPQVTPEAHTDFI) Monoclonal Antibody (03091YF) (CAT#: VRH-03091F)This mouse monoclonal antibody 03091YF is generated from premade hybridoma library, and specific for Equid alphaherpesvirus 4 ORF74 (AA 169-199): KKPPTLPRVHVKTPPPILVPQVTPEAHTDFI. For any additional requirments, please specify and submit an inquiry for assistance.
Catalog No. NC2235668
Add to Cart
Request a Quote