Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse anti-Feline Ig Mix Secondary Antibody

Catalog No. p-7244512
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-609 500 μg
PISA510356 1 mg
PISA510357 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-671-609 Supplier Invitrogen™ Supplier No. SA110642
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Secondary Antibody

SA1-10642 detects feline Immunoglobulin. Does not react with bovine, equine, canine, rabbit, sheep, porcine, donkey, guinea pig, goat or human. SA1-10642 has been successfully used in ELISA applications. The SA1-10642 immunogen is purified feline immunoglobulin.

Anti-Feline secondary antibodies are affinity-purified antibodies with well-characterized specificity for feline immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Feline Ig Mix
Applications ELISA
Classification Monoclonal
Clone FIG10-118.1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Host Species Mouse
Immunogen Purified feline immunoglobulin.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Feline
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Secondary Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.