Learn More
Immunology Consultants Mouse anti-Fluorescein Isothiocyanate (FITC) Antibody, 1.0 mg
SDP
Supplier:  Immunology Consultants MFTC65A
Encompass_Preferred
This mouse IgG is purified using salt fractionation procedures and Protein A chromatography. This antibody reacts with Fluorescein Isothiocyanate (FITC) as determined by ELISA, western blot, and lateral flow. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-196-2089