Learn More
Electron Microscopy Sciences Aurion Mouse-Anti-FITC, UltraSmall 1.5 ML
SDP
Supplier:  Electron Microscopy Sciences 25580
Encompass_Preferred
For the electron microscopic visualization of FITC labels used in light microscopic specimens. For retrieval of faded FITC in DNA/RNA probes, fatty acids, and proteins.
Particle size: UltraSmall
Catalog No. 50-274-25