Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Anti-Goat IgG Antibody [1G12H5], ProSci
SDP

Catalog No. 16595617 Shop All ProSci Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-617 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-617 Supplier ProSci LLC Supplier No. PM62790050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Control Antibody

Mouse Anti-Goat IgG Antibody specifically detects Mouse Anti-Goat IgG in Goat samples. It is validated for ELISA, Western Blot, Immunohistochemistry

Specifications

Antigen Mouse Anti-Goat IgG
Applications ELISA, Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 1G12H5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunoblotting: 1/5,000 - 1/50,000, ELISA: 1/5,000 - 1/50,000, Immunohistochemistry: 1/100 - 1/1,000
Formulation PBS pH 7.2, 1% BSA, 0.02% Thimerosal
Host Species Mouse
Immunogen Goat IgG
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity No cross reactivity with immunoglobulins from other species.
Target Species Goat
Content And Storage Antibody can be stored at +4°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Product Type Antibody
Form Liquid
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.