Learn More
Jackson Immuno Research Labs Peroxidase AffiniPure Mouse Anti-Goat IgG (H+L), 1ml
SDP
Supplier:  Jackson Immuno Research Labs 205035108
Encompass_Preferred
Peroxidase-AffiniPure Mouse Anti-Goat IgG (H+L) (min X Ms,Hu,Rb Sr Prot)
Catalog No. NC9838192