Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-HA-Tag, MP Biomedicals™

Catalog No. MP08L100032 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08L100032 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MP08L100032 Supplier MP Biomedicals, Inc Supplier No. 08L100032
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

  • Mouse anti HA-Tag mAb offers high specificity and potency and consistency against HA tag, which has been experimentally validated in Western blotting (WB), Immunoprecipitation (IP), and Immunofluorescence (IF) assays.
  • Protein tags are peptide sequences genetically grafted onto a recombinant protein. Often these tags are removable by chemical agents or by enzymatic means, such as proteolysis or intein splicing.
  • Tags are attached to proteins for various purposes. Epitope tags are short peptide sequences that are chosen because high-affinity antibodies can be reliably produced in many different species. These are usually derived from viral genes, which explain their high immunoreactivity. Epitope tags include V5-tag, Myc-tag, HA-tag, and NE-tag. These tags are particularly useful for western blotting, immunofluorescence, and immunoprecipitation experiments, although they also find use in antibody purification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HA-Tag
Applications Western Blot, Immunoprecipitation, Immunofluorescence
Classification Monoclonal
Concentration WB: 1:2000 - 1:5000; IF: 1:50 - 1:500; IP: 1:20 - 1:50
Conjugate Unconjugated
Formulation In PBS buffer with 0.02% sodium azide, 0.05% BSA, 50% glycerol; pH 7.3 to 7.4
Host Species Mouse
Immunogen Synthetic peptide corresponding to HA tag
Purification Method Affinity Purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Secondary
Antibody Molecular Weight Observed at 40 kDa
Form Purified
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.