Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Hamster IgG, Clone: G192-1, Secondary Antibody, BD

Catalog No. BDB554024 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554024 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554024 Supplier BD Biosciences Supplier No. 554024
Add to Cart
Add to Cart

Based on ELISA, the G192-1 antibody reacts specifically with Armenian hamster IgG2, IgG3, and IgG4, and Syrian hamster IgG2 monoclonal antibodies. The G192-1 mAb does not react with other hamster IgG groups or hamster IgM

Host Species: Mouse
Clone: G192-1
Isotype: IgG1, κ
Species Reactivity: Hamster
Immunogen: Pooled Armenian and Syrian hamster IgG mAb

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Hamster IgG
Classification Monoclonal
Clone G192-1
Concentration 0.5mg/mL
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Hamster
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.