Learn More
Mouse anti-Hamster IgG Cocktail, Clone: G94-90.5, G70-204, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
This preparation consists of a mixture of two mouse anti-hamster IgG monoclonal antibodies. It reacts with Armenian hamster IgG1/2/3 and Syrian hamster IgG1 monoclonal antibodies. It does not react with other hamster IgG groups and hamster IgM antibodies.
Host Species: Mouse
Clone: G94-90.5, G70-204
Isotype: IgG1, κ and IgG1, κ
Species Reactivity: Hamster
Immunogen: Pooled Armenian Hamster IgG mAb
Flow Cytometry
Specifications
Specifications
| Antigen | Mouse anti-Hamster IgG Cocktail |
| Classification | Monoclonal |
| Clone | G70-204, G94-90.5 |
| Conjugate | PE |
| Host Species | Mouse |
| Quantity | 0.2 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Target Species | Hamster |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.