Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Hamster IgG1, Clone: G94-56, Purified, Secondary Antibody, BD

Catalog No. BDB554006 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554006 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554006 Supplier BD Biosciences Supplier No. 554006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Based on ELISA, the G94-56 antibody reacts specifically with Armenian and Syrian hamster IgG1 monoclonal antibodies, but has low reactivity with some other hamster IgG groups. (Please refer to the Reactivity of Mouse Anti-Hamster Ig mAbs chart on our website for more information: http://www.bdbiosciences.com/pharmingen/hamster_chart_11x17.pdf.) The G94-56 mAb does not react with hamster IgM.

Host Species: Mouse
Clone: G94-56
Isotype: IgG2b, κ
Species Reactivity: Hamster
Immunogen: Pooled Armenian hamster IgG mAb

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Hamster IgG1
Classification Monoclonal
Clone G94-56
Concentration 0.5mg/mL
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Hamster
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.