Learn More
Mouse anti-Hamster IgG1, Clone: G94-56, Biotin, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
Based on ELISA, the G94-56 antibody reacts specifically with Armenian and Syrian hamster IgG1 monoclonal antibodies, but has low reactivity with some other hamster IgG groups. (Please refer to the Reactivity of Mouse Anti-Hamster Ig mAbs chart on our website for more information: http://www.bdbiosciences.com/pharmingen/hamster_chart_11x17.pdf.) The G94-56 mAb does not react with hamster IgM.
Host Species: Mouse
Clone: G94-56
Isotype: IgG2b, κ
Species Reactivity: Hamster
Immunogen: Pooled Armenian hamster IgG mAb
ELISA, Flow Cytometry
Specifications
Specifications
| Antigen | Mouse anti-Hamster IgG1 |
| Classification | Monoclonal |
| Clone | G94-56 |
| Concentration | 0.5mg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.