Learn More
Mouse anti-Hamster IgM, Clone: G188-2, FITC, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
Based on ELISA, the G188-2 antibody reacts specifically with hamster IgM. The G188-2 mAb does not react with hamster IgG
Host Species: Mouse
Clone: G188-2
Isotype: IgG1, κ
Species Reactivity: Hamster
Immunogen: Armenian Hamster IgM (mAb HM40-3)
Flow Cytometry
Specifications
Specifications
| Antigen | Mouse anti-Hamster IgM |
| Classification | Monoclonal |
| Clone | G188-2 |
| Concentration | 0.5mg/mL |
| Conjugate | FITC |
| Host Species | Mouse |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.