Learn More
Chondrex Inc MOUSE ANTI-HDM IGE ASSAY KT
Catalog No. 501526766
Encompass_Preferred
ELISA kit for assaying mouse anti-house dust mite (HDM) IgE antibodies in mouse serum and plasma samples. Kit contains a 96-well ELISA plate pre-coated with HDM extract from Dermatophagoides pteronyssinus, as well as all reagents needed to complete the assay. Standard Range: 100 ng/mL to 1.6 ng/mL. Working Time: 4.5 hours
Catalog No. 50-152-6766 Supplier Chondrex Inc Supplier No. 3037
ELISA kit for assaying mouse anti-house dust mite (HDM) IgE antibodies in mouse serum and plasma samples. Kit contains a 96-well ELISA plate pre-coated with HDM extract from Dermatophagoides pteronyssinus, as well as all reagents needed to complete the assay. Standard Range: 100 ng/mL to 1.6 ng/mL. Working Time: 4.5 hours