Learn More
Chondrex Inc Mouse Anti-House Dust Mite (HDM) IgG Antibody Assay Kit
Supplier:  Chondrex Inc 3030
Encompass_Preferred
Regarding anti-HDM IgG levels, anti-HDM IgG1 antibodies are associated with Th2 responses, while anti-HDM IgG2a or IgG2b antibodies are associated with Th1 responses. Therefore, measuring the antibody levels of anti-HDM IgE and IgG can be useful for determining effective sensitization and treatments. ASSAY TIME: 4.5 hours, STANDARD RANGE: 100 - 1.6 ng/ml, NUMBER OF SAMPLES: Up to 40 (duplicate) samples/plate, SAMPLE TYPES: Serum & Plasma (pre-treatment acceptable)
Catalog No. NC1929621