Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human Ig κ Light Chain, Clone: G20-361, Secondary Antibody, BD

Catalog No. BDB555861 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555861 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555861 Supplier BD Biosciences Supplier No. 555861
Add to Cart
Add to Cart

The G20-361 antibody reacts with human immunoglobulin light chain kappa (κ). It does not react with λ light chain or heavy chains. This antibody is routinely tested by ELISA analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: G20-361
Isotype: IgG2b, κ
Species Reactivity: Human

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Human Ig κ Light Chain
Classification Monoclonal
Clone G20-361
Concentration 0.5mg/mL
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Human
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.