Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Human IgA Secretory Mouse anti-Human, Clone: 4G10.1, Secondary Antibody, MilliporeSigma™

Catalog No. MAC132MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
400 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAC132MI 400 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAC132MI Supplier MilliporeSigma Supplier No. MAC132
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Mouse Anti-Human IgA, Secretory Antibody, Clone 4G10.1 is validated for use in Enzyme Immunoassay (ELISA) for the detection of Mouse Human IgA, Secretory.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgA
Applications ELISA
Classification Monoclonal
Clone 4G10.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing PBS without preservatives.
Host Species Mouse
Immunogen Purified antibody corresponding to Human IgA, Secretory.
Purification Method Protein G purified
Quantity 400 μg
Primary or Secondary Secondary
Target Species Human
Content And Storage −20°C for up to 24 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.