Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgA Secondary Antibody, HRP, Invitrogen™

Catalog No. 01672140
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-140 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-140 Supplier Thermo Scientific Supplier No. SA135467

Mouse Monoclonal Secondary Antibody

This antibody does not cross react with albumin fractions of human serum, human IgG, human IgM or albumin fractions from other species.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgA
Applications Dot Blot, ELISA, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone BFG-267
Concentration 1.2 mg/mL
Conjugate HRP
Formulation PBS with 50% glycerol, 5mg/mL BSA and no preservative; pH 7.5
Host Species Mouse
Immunogen Purified alpha-chain (Fc) of human IgA
Purification Method Ion-exchange chromatography, Size-exclusion - Gel filtration
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage -20°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.