Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgD, Clone: IA6-2, PE, Secondary Antibody, BD

Catalog No. BDB555779 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555779 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555779 Supplier BD Biosciences Supplier No. 555779
Add to Cart
Add to Cart

Reacts with the heavy chain of human IgD expressed on B cells. This reactivity is neutralized by purified human IgD and not by purified human IgM. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: IA6-2
Isotype: IgG2a, κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Human IgD
Classification Monoclonal
Clone IA6-2
Conjugate PE
Host Species Mouse
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Human
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.