Learn More
Mouse anti-Human IgE, Clone: G7-26, biotin, Secondary Antibody, BD
Shop All Waters Corporation ProductsDescription
The G7-26 antibody reacts with human immunoglobulin E. It does not react with other heavy chain isotypes.
Host Species: Mouse
Clone: G7-26
Isotype: IgG2a, κ
Species Reactivity: Human
ELISA
Specifications
Specifications
| Antigen | Mouse anti-Human IgE |
| Classification | Monoclonal |
| Clone | G7-26 |
| Concentration | 0.5mg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.