Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgE, Clone: G7-26, Secondary Antibody, BD

Catalog No. BDB555857 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555857 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555857 Supplier BD Biosciences Supplier No. 555857
Add to Cart
Add to Cart

The G7-26 antibody reacts with human immunoglobulin E. It does not react with other heavy chain isotypes.

Host Species: Mouse
Clone: G7-26
Isotype: IgG2a, κ
Species Reactivity: Human

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Human IgE
Classification Monoclonal
Clone G7-26
Concentration 0.5mg/mL
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Human
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.