Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG, Clone: G18-145, PE, Secondary Antibody, BD

Catalog No. BDB555787 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555787 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555787 Supplier BD Biosciences Supplier No. 555787
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reacts with the heavy chain of human immunoglobulin subclasses: IgG1, IgG2, IgG3 and IgG4. Does not react with other heavy chain isotypes. This antibody is routinely tested by ELISA analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: G18-145
Isotype: IgG1, κ
Species Reactivity: Human

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Human IgG
Classification Monoclonal
Clone G18-145
Conjugate PE
Host Species Mouse
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Human
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.