Learn More
Mouse anti-Human IgG F(ab) Secondary Antibody (4A11), DyLight 350, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgG F(ab) |
| Applications | ELISA, Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 4A11 |
| Conjugate | DyLight 350 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Purified human IgG. |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.