Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG F(ab) Secondary Antibody (4A11), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB535007
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB535007 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB535007 Supplier R&D Systems Supplier No. NB500467FR

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

IgG F(ab) Monoclonal specifically detects IgG F(ab) in Human samples. It is validated for ELISA, Flow Cytometry, Western Blot.

Specifications

Antigen IgG F(ab)
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4A11
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Purified human IgG.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.