Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG F(ab) Secondary Antibody (4A11), PE, Novus Biologicals™
SDP

Catalog No. NBP144932
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144932 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144932 Supplier R&D Systems Supplier No. NBP144932

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

IgG F(ab) Monoclonal specifically detects IgG F(ab) in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IgG F(ab)
Applications Flow Cytometry
Classification Monoclonal
Clone 4A11
Concentration 0.1 mg/mL
Conjugate PE
Dilution Western Blot, Flow Cytometry 1-5 μg/mL, ELISA
Gene Alias immunoglobulin heavy constant gamma 1 (G1m marker)
Gene Symbols IGHG1
Host Species Mouse
Immunogen Purified human IgG.
Purification Method Size Exclusion Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity The antibody 4A11 reacts with Fab fragment of human IgG.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.