Learn More
Genscript Corporation Mouse Anti-Human IgG Fab Antibody (12H3C4A6)[HRP], mAb
Supplier:  Genscript Corporation A01855200
Encompass
GenScript Mouse Anti-Human IgG Fab Antibody (12H3C4A6)[HRP], mAb reacts with the Fab portion of human IgG but not with the Fc portion of human IgG. The monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken or goat immunoglobulins.
Catalog No. 50-227-1975