Learn More
Medchemexpress LLC Mouse Anti-Human IgG Fc | 1 MG
SDP
Catalog No. 5000443357
Encompass_Preferred
Mouse Anti-Human IgG Fc is a mouse-derived and non-conjugated IgG2a recombinant monoclonal antibody, targeting to IgG Fc. It is expressed from CHO.
  • Mouse-derived
  • Non-conjugated IgG2a recombinant monoclonal antibody
  • Targets IgG Fc
  • Expressed from CHO
  • Heavy chain type is mIg2a
  • Light chain type is mkappa
Catalog No. 50-004-43357 Supplier Medchemexpress LLC Supplier No. HYP837271MG
Mouse Anti-Human IgG Fc is a mouse-derived and non-conjugated IgG2a recombinant monoclonal antibody, targeting to IgG Fc. It is expressed from CHO.
  • Mouse-derived
  • Non-conjugated IgG2a recombinant monoclonal antibody
  • Targets IgG Fc
  • Expressed from CHO
  • Heavy chain type is mIg2a
  • Light chain type is mkappa