Learn More
Immunology Consultants anti-Human IgG Fc Antibody (Mouse Clone #907)
SDP
Catalog No. 502248893
Encompass_Preferred
This monoclonal antibody provides sensitive, specific detection of the human IgG Fc region, supporting ELISA, Western blotting, and immunoassay development requiring Fc‑directed recognition. Clone 907 delivers strong affinity and low nonspecific binding, enabling accurate measurement of human IgG in serum, plasma, and purified antibody preparations. It is ideal for therapeutic antibody characterization, immunoassay calibration, and studies examining Fc‑mediated immune interactions. MGFC‑80A‑907 offers consistent lot‑to‑lot performance, ensuring reliable Fc detection across diverse analytical applications.
This item is not returnable. View return policy
Catalog No. 50-224-8893 Supplier Immunology Consultants Supplier No. MGFC80A907
This monoclonal antibody provides sensitive, specific detection of the human IgG Fc region, supporting ELISA, Western blotting, and immunoassay development requiring Fc‑directed recognition. Clone 907 delivers strong affinity and low nonspecific binding, enabling accurate measurement of human IgG in serum, plasma, and purified antibody preparations. It is ideal for therapeutic antibody characterization, immunoassay calibration, and studies examining Fc‑mediated immune interactions. MGFC‑80A‑907 offers consistent lot‑to‑lot performance, ensuring reliable Fc detection across diverse analytical applications.