Learn More
Genscript Corporation Mouse Anti-Human IgG Fc Antibo
Catalog No. 5000058357
Encompass
GenScript Mouse Anti-Human IgG Fc Antibody (50B4A9) mAb reacts with the Fc portion of human IgG but not with the Fab portion of human IgG This monoclonal antibody has no cross-reactivity with mouse rat rabbit chicken or goat immunoglobulins
Catalog No. 50-000-58357 Supplier Genscript Corporation Supplier No. A02257200
GenScript Mouse Anti-Human IgG Fc Antibody (50B4A9) mAb reacts with the Fc portion of human IgG but not with the Fab portion of human IgG This monoclonal antibody has no cross-reactivity with mouse rat rabbit chicken or goat immunoglobulins